Description
The Mmp9 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of mouse Mmp9. This antibody recognizes mouse antigen. The Mmp9 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
Specifications
| Antigen | Mmp9 |
| Applications | Western Blot, ELISA |
| Clone | 5A1 |
| Concentration | 1000 μg/mL |
| Gene Symbols | AW743869, B/MMP9, Clg4b, matrix metallopeptidase 9, MMP 9, Mmp9, pro MMP 9 |
| Host Species | Rabbit |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 17395 |
| Target Species | Mouse |
| Show More |