missing translation for 'onlineSavingsMsg'
Learn More

MMP-14/MT1-MMP Antibody (3G4.2) - BSA Free, Novus Biologicals™

Product Code. p-7200785 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
Product Code. Quantity unitSize
18120404 0.025 mg 25µg
18153633 0.1 mg 0.1mg
2 options
This item is not returnable. View return policy

Product Code. 18120404

Brand: Novus Biologicals NBP2297350.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMP-14/MT1-MMP Monoclonal specifically detects MMP-14/MT1-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-14/MT1-MMP
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 3G4.2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500
Formulation PBS (pH 7.2) with 0.1% Sodium Azide
Gene Accession No. P50281
Gene Alias EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1
Gene Symbols MMP14
Host Species Mouse
Immunogen Recombinant protein containing the catalytic domain of human MT1-MMP.
Molecular Weight of Antigen 60 kDa
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4323
Test Specificity Clone 3G4.2 recognizes the catalytic domain of MT1-MMP, identifying the active form of the protein. MW is approximately 60 kDa.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.