Learn More
MMAF-d8 hydrochloride, MedChemExpress
Product Code. 16449207
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. Quantity unitSize
16449207 5 mg 5mg
16471529 1 mg 1mg
16436648 10 mg 10mg
3 options
Product Code. 16449207
Brand: Medchem Express HY15579AS
MedChemExpress D8-MMAF hydrochloride is a deuterated form of MMAF hydrochloride, which is a microtubule disrupting agent.
D8-MMAF hydrochloride is a deuterated form of MMAF hydrochloride, which is a microtubule disrupting agent.
Specifications
Chemical Name or Material MMAF-d8 hydrochloride
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C39H58D8ClN5O8
Technique Cancer-programmed Cell Death
Color Off-White
Solubility Information DMSO : 100 mg/mL (128.79 mM; Need ultrasonic)
SMILES CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C(O)=O)CC2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C.Cl
Molecular Weight (g/mol) 776.47
Quantity 5 mg
Formula Weight 776.47
Percent Purity 98.97%
Grade Research