Learn More
MMAF-d8 hydrochloride, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
D8-MMAF hydrochloride is a deuterated form of MMAF hydrochloride, which is a microtubule disrupting agent.
Specifications
Specifications
| Chemical Name or Material | MMAF-d8 hydrochloride |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C39H58D8ClN5O8 |
| Technique | Cancer-programmed Cell Death |
| Color | Off-White |
| Solubility Information | DMSO : 100 mg/mL (128.79 mM; Need ultrasonic) |
| SMILES | CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C(O)=O)CC2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C.Cl |
| Molecular Weight (g/mol) | 776.47 |
| Quantity | 10 mg |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.