Learn More
MMAE-d8, MedChemExpress
Product Code. 16476338
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. 16476338
Brand: Medchem Express HY15162A
MedChemExpress MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.
MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.
Specifications
Chemical Name or Material MMAE-d8
CAS 2070009-72-0
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C39H59D8N5O7
Technique Cancer-programmed Cell Death
Color White
Synonym Monomethyl auristatin E-d8 Deuterated labeled MMAE
Solubility Information DMSO : 100 mg/mL (137.74 mM; Need ultrasonic)
SMILES CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C)[C@@H](O)C2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C(C([2H])([2H])[2H])(C([2H])([2H])[2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C
Molecular Weight (g/mol) 726.03
Quantity 10 mg
Formula Weight 726.03
Percent Purity 98.43%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.