missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11996-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
MAP3K11 is a member of the serine/threonine kinase family. This kinase contains a SH3 domain and a leucine zipper-basic motif. This kinase preferentially activates MAPK8/JNK kinase, and functions as a positive regulator of JNK signaling pathway. This kinase can directly phosphorylate, and activates IkappaB kinase alpha and beta, and is found to be involved in the transcription activity of NF-kappaB mediated by Rho family GTPases and CDC42.Specifications
MLK3 | |
Polyclonal | |
Unconjugated | |
MAP3K11 | |
MAP3K11, Mixed lineage kinase 3, MLK 3, MLK3, PTK1, SPRK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26403, 309168, 4296 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16584, Q66HA1, Q80XI6 | |
MAP3K11 | |
MLK3 Fusion Protein Ag2636 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |