missing translation for 'onlineSavingsMsg'
Learn More

MLF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11835-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889163

  • 3468.15 NOK / 150µL
Estimated Shipment: 19-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MLF2 is a protein coding gene.
Specifications

Specifications

MLF2
Polyclonal
Unconjugated
MLF2
MLF2, myeloid leukemia factor 2, NTN4
Rabbit
Antigen Affinity Chromatography
RUO
30853, 312709, 8079
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15773, Q99KX1
Mlf2
MLF2 Fusion Protein Ag2270
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers