Learn More
VistaLab™ Technologies MLA™ D-Tipper™ Pipetters
GREENER_CHOICE
Product Code. 11985802
Change view
Click to view available options
Volume (Metric):
10 μL
10, 50, 100, 200, 1000 μL
100 μL
1000 μL
120 μL
15 μL
150 μL
20 μL
200 μL
25 μL
250 μL
30 μL
300 μL
40 μL
400 μL
5 μL
50 μL
500 μL
60 μL
600 μL
70 μL
700 μL
75 μL
750 μL
80 μL
90 μL
900 μL
Unit Size:
1 set
Each
Product Code. Volume (Metric) unitSize
11985802 300 μL Each
11985882 600 μL Each
11995892 700 μL Each
11975882 750 μL Each
11925862 15 μL Each
11945862 40 μL Each
11965862 60 μL Each
11975862 70 μL Each
11985862 75 μL Each
11995862 80 μL Each
11905872 90 μL Each
11915872 120 μL Each
11925872 150 μL Each
11985892 900 μL Each
11935862 30 μL Each
11935872 400 μL Each
11955792 25 μL Each
11995792 200 μL Each
11975802 250 μL Each
11935792 10 μL Each
11945792 20 μL Each
11965792 50 μL Each
11975792 100 μL Each
11995802 500 μL Each
11985812 1000 μL Each
11915862 5 μL Each
11935942 10, 50, 100, 200, 1000 μL 1 set
27 options
Product Code. 11985802
Brand: VistaLab™ Technologies 1058C
Only null left
Add to basket
Add to basket
Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Color coded body for easy volume identification and individually serialized with Calibration Certificate included.
  • Single-stroke 'to deliver' operation
  • Built-in tip-ejector and glove guard
  • Factory calibrated to deliver specific quantities; can be easily recalibrated in the lab
  • Available in 30 fixed volumes ranging from 5-1000 μL
  • Also available as a Selectable model: 50/100/200 μL
Specifications
Accuracy ±1%
Autoclavable Not Autoclavable
Color Brown
Disposable Reusable
Material Aluminum, Stainless Steel
Model D-Tipper
Number of Channels 1
Sterility Non-sterile
Type Axial Pipette
Volume (Metric) 300 μL
For Use With (Application) Micro Pipetting/Liquid Transfer
Includes Nozzle Insert, Calibration Key
Precision 0.60%
Warranty Limited Lifetime