Learn More
ML277, MedChemExpress
Product Code. 16475108
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16475108
Brand: Medchem Express HY12343
MedChemExpress ML277(CID53347902) is a novel, potent and selective K(v)7.1 (KCNQ1) potassium channel activator with EC50 of 270 nM.
ML277(CID53347902) is a novel, potent and selective K(v)7.1 (KCNQ1) potassium channel activator with EC50 of 270 nM.
Specifications
Chemical Name or Material ML277
CAS 1401242-74-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H25N3O4S2
Quantity 10 mg
Synonym CID-53347902
Solubility Information DMSO : ≥ 56 mg/mL (118.75 mM)
SMILES O=C([C@@H]1N(S(=O)(C2=CC=C(C)C=C2)=O)CCCC1)NC3=NC(C4=CC=C(OC)C=C4)=CS3
Molecular Weight (g/mol) 471.59
Formula Weight 471.59
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.