Learn More
ML264, MedChemExpress
Product Code. 16490468
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16490468 5 mg 5mg
16458008 10 mg 10mg
16447786 100 mg 100mg
16413937 50 mg 50mg
4 options
Product Code. 16490468
Brand: Medchem Express HY19994
MedChemExpress ML264 is an antitumor agent that potently and selectively inhibits Krüppel-like factor five (KLF5) expression.
ML264 is an antitumor agent that potently and selectively inhibits Krüppel-like factor five (KLF5) expression.
Specifications
Chemical Name or Material ML264
Physical Form Solid
CAS 1550008-55-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H21ClN2O4S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 100 mg/mL (259.82 mM)
SMILES O=C(NCC(N(C)C(CC1)CCS1(=O)=O)=O)/C=C/C2=CC=CC(Cl)=C2
Molecular Weight (g/mol) 384.88
Formula Weight 384.88
Percent Purity 99.58%
Grade Research