missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKC8866, MedChemExpress
Description
MKC8866 strongly inhibits Dithiothreitol-induced X-box-binding protein 1-spliced (XBP1s) expression with an EC50 of 0.52 μM and unstresses RPMI 8226 cells with an IC50 of 0.14 μM. MKC8866 inhibits IRE1 RNase in breast cancer cells leading to the decreased production of pro-tumorigenic factors and it can inhibits prostate cancer (PCa) tumor growth.
Specifications
Specifications
| Chemical Name or Material | MKC8866 |
| CAS | 1338934-59-0 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C18H19NO7 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 16.67 mg/mL (46.13 mM; Need ultrasonic) |
| SMILES | O=CC1=C(O2)C(C(C)=C(CC(N3CCOCC3)=O)C2=O)=CC(OC)=C1O |
| Molecular Weight (g/mol) | 361.35 |
| Formula Weight | 361.35 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?