Learn More
MK-6892, MedChemExpress
Product Code. 16456797
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16456797
Brand: Medchem Express HY10680
MedChemExpress MK-6892 is a potent, selective, and full agonist for the high affinity nicotinic acid (NA) receptor GPR109A. Ki and GTPγS EC50 of MK-6892 on the Human GPR109A is 4 nM and 16 nM, respectively.
MK-6892 is a potent, selective, and full agonist for the high affinity nicotinic acid (NA) receptor GPR109A. Ki and GTPγS EC50 of MK-6892 on the Human GPR109A is 4 nM and 16 nM, respectively.
Specifications
Chemical Name or Material MK-6892
CAS 917910-45-3
Color White
Purity Grade Notes Research
Molecular Formula C19H22N4O5
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (129.40 mM; Need ultrasonic)
SMILES OC(C=C1)=CN=C1C2=NOC(CC(C)(C)C(NC3=C(C(O)=O)CCCC3)=O)=N2
Molecular Weight (g/mol) 386.4
Formula Weight 386.4
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.