missing translation for 'onlineSavingsMsg'
Learn More

MK-2461, MedChemExpress

Product Code. p-200098452
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
Product Code. Quantity unitSize
16478088 10 mg 10mg
16463756 25 mg 25mg
16400558 5 mg 5mg
16400356 2 mg 2mg
16464007 50 mg 50mg
5 options
This item is not returnable. View return policy

Product Code. 16478088

Brand: Medchem Express HY50703

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-2461 is a novel ATP-competitive multitargeted inhibitor of activated c-Met with a mean IC50 of 2.5 nM.

MK-2461 is a novel ATP-competitive multitargeted inhibitor of activated c-Met with a mean IC50 of 2.5 nM.

Specifications

Chemical Name or Material MK-2461
Physical Form Solid
CAS 917879-39-1
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H25N5O5S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 31 mg/mL (62.56 mM)
SMILES O=S(N(C)C[C@H]1OCCOC1)(NC2=CC=C3C=CC(C(C(C3=C2)=O)=C4)=NC=C4C5=CN(N=C5)C)=O
Molecular Weight (g/mol) 495.55
Formula Weight 495.55
Percent Purity 99.01%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.