missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MK-2206 dihydrochloride, MedChemExpress
Description
MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy.
Specifications
Specifications
| Chemical Name or Material | MK-2206 dihydrochloride |
| CAS | 1032350-13-2 |
| Color | Green Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C25H23Cl2N5O |
| Quantity | 10 mg |
| Synonym | MK-2206 (2HCl) |
| Solubility Information | DMSO : 16.67 mg/mL (34.70 mM; ultrasonic and warming and heat to 70°C) ∣H2O : 3.57 mg/mL (7.43 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C1N2C(C3=CC(C4=CC=CC=C4)=C(N=C3C=C2)C5=CC=C(C6(N)CCC6)C=C5)=NN1.Cl.Cl |
| Molecular Weight (g/mol) | 480.39 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?