Learn More
Hirschmann™ Class A Measuring Cylinders with NS Ground and Glass Stopper
Product Code. p-8013434
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Graduations:
0.2 mL
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Pack of 2
Product Code. 10141481
Brand: Hirschmann™ 2340293
Only null left
Add to basket
Add to basket
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann™ Class A Measuring Cylinders with NS Ground and Glass Stopper are manufactured from durable DURAN™ glass tubing.
  • Blue, main point graduation
  • With NS ground and glass stopper
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Base Type Hexagonal
Scale Color Blue
Shape Round
Type Graduated Mixing Cylinder
Tolerance ±5 mL
Material Glass
Certifications/Compliance DIN EN ISO 4788
Class A
Capacity (Metric) 1000 mL
Height (Metric) 510 mm
Includes Glass Stopper
Stopper Material Glass
Graduations 10 mL