Learn More
Kinematica™ Mixing Attachment for Microtron™ MB 550 Laboratory Mixer
Shop All Kinematica Products
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Description
- For use with the MB 550 Mixers
- The two stainless steel ball bearings are sealed twice each
- Noiseless and sturdy coupling with dampers
- Blades, sealing and dampers can easily be exchanged
- Four high-grade blades designed for optimal mixing
- Generally suited for intensive and fast processing
Order Info
dimCapacityMetric
Specifications
Specifications
| Product Type | Mixing Head Attachment |
| Capacity (Metric) | 1000 mL |
| Blade Material | Stainless Steel |
| Material | Borosilicate Glass |
| Description | Consists of gasket and blade head |
| For Use With (Equipment) | MB 550 Laboratory Mixer |
| Packaging Quantity | 1 Ea. |