Learn More
Cytiva Mixed Cellulose Ester Filtration Membrane
Product Code. 12465242
Click to view available options
Unit Size:
Pack of 100
Product Code. 12465242
Brand: Cytiva 10401664
These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters
Whatman mixed cellulose ester membranes are composed of cellulose acetate and cellulose nitrate. These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters. Also, the color contrast provided by the filter surface facilitates particle detection and minimises eye fatigue. The ME range has a lower cellulose acetate content compared to the WME range of membranes. Circles, gridded.
  • Sterile options available for critical applications
  • Excellent contrast for easier particle detection
  • Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
  • Black plain and black gridded membranes have a mix of cellulose nitrate and cellulose acetate
  • The membrane offers a high degree of internal surface area for greater adsorption of product
  • Higher dirt loading capacity
  • Biologically inert with good thermal stability
  • No surfactants to contaminate samples
  • Uniform microporous structure of membrane gives high flow rates
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material MCE
Pore Size 0.45 μm
Sterility Non-sterile
Flow Rate 12.5/100/12.5sec./mL/cm2
For Use With (Application) Microbiological Colony Counting Procedures
Grid Color Plain
Membrane Type ME25
No. per Pack 100Pack
Type Membrane Filter
compliance-icons
Signal Word
  • ||Warning
Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose