Learn More
Macherey-Nagel™ Mix IV Diamino/Carbon Clean-up Mix
Product Code. 12891031 Shop All Macherey-Nagel Products
Change view
Click to view available options
Volume (Metric):
15 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12891031 15 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12891031 Supplier Macherey-Nagel™ Supplier No. 730973
  • ONLINE EXCLUSIVE PRICE
    1675.00 NOK
    1534.30 NOK / Pack of 1
    Save 140.70 NOK
    8% Off
In Stock
Specifications
For Use With (Equipment) Chromabond QuEChERS Clean-up Mixes (EN 15662)
Type Diamino/Carbon Clean-up Mix
Volume (Metric) 15 mL
Quantity 50 Pack