missing translation for 'onlineSavingsMsg'
Learn More

Mitochonic acid 5, MedChemExpress

Product Code. p-200100725
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490797

Brand: Medchem Express HY111536

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mitochonic acid 5 binds mitochondria and ameliorates renal tubular and cardiac myocyte damage. Mitochonic acid 5 modulates mitochondrial ATP synthesis.

Mitochonic acid 5 binds mitochondria and ameliorates renal tubular and cardiac myocyte damage. Mitochonic acid 5 modulates mitochondrial ATP synthesis.

Specifications

Chemical Name or Material Mitochonic acid 5
CAS 1354707-41-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H13F2NO3
Quantity 50 mg
Synonym MA-5
Solubility Information DMSO : ≥ 106.66 mg/mL (323.90 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)C(CC(C1=CC=C(F)C=C1F)=O)C2=CNC3=C2C=CC=C3
Molecular Weight (g/mol) 329.3
Formula Weight 329.3
Percent Purity 99.28%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.