Learn More
MIS12 Mouse anti-Human, Unconjugated, Clone: 2G5F6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67475-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16890116
  • 4593.76 NOK / 150µL
Estimated Shipment: 17-04-2024
Description
The MIS12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MIS12. This antibody recognizes human antigen. The MIS12 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
MIS12
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
2G5F6
Unconjugated
150 μL
79003
IgG2a