Learn More
Mirtazapine-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Mirtazapine-d3 |
| CAS | 1216678-68-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 2H3 H16 N3 |
| Quantity | 1 mg |
| Synonym | Mirtazapine-D3 |
| InChI Formula | InChI=1S/C17H19N3/c1-19-9-10-20-16(12-19)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)20/h2-8,16H,9-12H2,1H3/i1D3 |
| SMILES | [2H]C([2H])([2H])N1CCN2C(C1)c3ccccc3Cc4cccnc24 |
| Molecular Weight (g/mol) | 268.37 |
| Formula Weight | 268.1767 |
| Show More |