Learn More
Minoxidil, Mikromol™
Description
Minoxidil
Specifications
Specifications
| Chemical Name or Material | Minoxidil |
| CAS | 38304-91-5 |
| Analyte or Component Names | Minoxidil |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H15 N5 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2,4-Diamino-6-piperidinopyrimidine 3-N-oxide, 2,4-Diamino-6-piperidinopyrimidine 3-oxide, 6-Amino-1,2-dihydro-1-hydroxy-2-imino-4-piperidinopyrimidine, Alopexil, Alostil, Loniten, Lonolox, Minoxidil, Minoximen, Mintop, Normoxidil, PDP, Pierminox, Prexidil, RiUP, Rogaine, Tricoxidil, U 10858 |
| InChI Formula | InChI=1S/C9H15N5O/c10-7-6-8(12-9(11)14(7)15)13-4-2-1-3-5-13/h6H,1-5,10H2,(H2,11,12) |
| SMILES | Nc1cc(nc(N)[n+]1[O-])N2CCCCC2 |
| IUPAC Name | 3-oxido-6-piperidin-1-ylpyrimidin-3-ium-2,4-diamine |
| Show More |