missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MINK1, GST Tag Recombinant Protein
MINK1 is a serine/threonine kinase belonging to the germinal center kinase (GCK) family.
16000.00 NOK - 69300.00 NOK
Specifications
Accession Number | NP_056531 |
---|---|
Molecular Weight (g/mol) | 63.9 kDa |
Gene Alias | MINK1, MINK |
KinaseFamily | STE20 Kinase Family |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478533
|
Invitrogen™
PR7198A |
100μg |
16000.00 NOK
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10757173
|
Invitrogen™
PV4022 |
1mg |
69300.00 NOK
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_056531 | |
MINK1, MINK | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: STE Group | |
Baculovirus |
63.9 kDa | |
STE20 Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice |