Learn More
Cytiva Minimate™ Tangential Flow Filtration (TFF) Capsule with Omega™ Membrane
Description
- Efficient processing
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
| Material (Membrane) | PES |
| MWCO | 5 kDa |
| Sterility | Non-sterile |
| Filtration Area | 50 cm2 |
| No. per Pack | 1/Pk. |