Learn More
Cytiva Minimate; EVO Tangential Flow Filtration (TFF) System
Product Code. 17184771
Change view
Click to view available options
Volume (Metric):
<500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17184771 <500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17184771 Supplier Cytiva Supplier No. OAPMPUNV
  • ONLINE EXCLUSIVE PRICE
    75820.00 NOK
    70229.64 NOK / Pack of 1
    Save 5590.36 NOK
    7% Off
Estimated Shipment: 28-12-2026
Plug-and-play system to streamline laboratory-scale concentration, desalting and buffer exchange processes. System includes all the hardware, tubing and fittings needed to get your process up and running quickly.
The low system working volume achieved through the use of a conical bottom reservoir and compact design enables high concentration factors from up to 1 L or more of sample to be achieved. Concentrate your sample down to as little as 5 mL. Designed to work with Minimate TFF capsules.
Roller head peristaltic pumps provide gentle processing and are the choice for critical applications such as fragile biomolecules.
System components are designed for easy assembly and disassembly; no tools required. Tubing and fitting replacement is simple and quick. Slip nuts for luer locks eliminate tubing kinks.
Applications:
  • Concentrate and desalt proteins, peptides, or nucleic acids (DNA, RNA, oligonucleotides)
  • Recover antibodies or recombinant proteins from clarified cell culture media
  • Process metal sensitive enzymes and molecules
  • Separate (fractionate) large from small biomolecules
  • Concentrate viruses or gene therapy vectors
  • Prepare samples prior to column chromatography
  • Concentrate samples after gel filtration
  • Depyrogenate water, buffers, and media solutions
TRUSTED_SUSTAINABILITY
Specifications
Description Non-sterile
Material Polysulfone
Volume (Metric) <500 mL
Dimensions (L x W x H) 48.2 x 30.7 x 20.8 cm