Learn More
Bryson Packaging™ Minigrip™ Polyethylene Write-on Bags
Product Code. 11998267
Change view
Click to view available options
Width (Metric):
57 mm
60 mm
90 mm
115 mm
125 mm
150 mm
230 mm
254 mm
279 mm
Height (Metric):
60 mm
76 mm
115 mm
190 mm
230 mm
325 mm
356 mm
406 mm
Unit Size:
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) Unit Size
11998267 230 mm 325 mm Pack of 100
11388193 115 mm 115 mm Pack of 100
11368203 254 mm 356 mm Pack of 100
11378203 279 mm 406 mm Pack of 100
11358203 57 mm 76 mm Pack of 100
11378193 60 mm 60 mm Pack of 100
11587863 90 mm 115 mm Pack of 100
11597863 125 mm 190 mm Pack of 100
11988267 150 mm 230 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11998267 Supplier Bryson Packaging™ Supplier No. BAJ340130G
  • ONLINE EXCLUSIVE PRICE
    264.60 NOK
    111.87 NOK / Pack of 100
    Save 152.73 NOK
    57% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Store and send samples and stationary in resealable bags. Bryson Packaging™ Minigrip™ Polyethylene Write-on Bags are clear polyethylene bags with three white bars for labelling. Square and rectangular bags are available in a range of sizes. 100 and 1,000 packs.
  • Clear polyethylene bags with Grip Seal resealable strips
  • Three generously sized write-on labelling bars for easy identification and organization
  • Ideal for storing stationery and sending samples
  • Available in a range of sizes to meet storage needs
Specifications
Description Resealable Write-On Bag
Color Clear with White Bars
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Quantity 100 Pack
Height (Metric) 325 mm
Width (Metric) 230 mm
Thickness (Metric) 50 μm