Learn More
Bryson Packaging™ Polyethylene Bags
Product Code. 12300549
Change view
Click to view available options
Width (Metric):
40 mm
60 mm
75 mm
90 mm
100 mm
115 mm
125 mm
150 mm
190 mm
230 mm
255 mm
330 mm
Height (Metric):
65 mm
75 mm
80 mm
115 mm
140 mm
175 mm
190 mm
230 mm
325 mm
355 mm
457 mm
Color:
Clear
Clear with White Bars
Unit Size:
Pack of 100
Product Code. Width (Metric) Height (Metric) Color unitSize
12300549 190 mm 190 mm Clear Pack of 100
11398203 40 mm 65 mm Clear Pack of 100
11318213 60 mm 75 mm Clear Pack of 100
11328213 75 mm 80 mm Clear Pack of 100
11978267 90 mm 115 mm Clear Pack of 100
10673995 100 mm 140 mm Clear Pack of 100
11348213 115 mm 115 mm Clear Pack of 100
12320499 125 mm 175 mm Clear Pack of 100
10031474 125 mm 190 mm Clear Pack of 100
10472624 150 mm 230 mm Clear Pack of 100
11356934 230 mm 325 mm Clear Pack of 100
11332284 255 mm 355 mm Clear Pack of 100
11388203 330 mm 457 mm Clear with White Bars Pack of 100
10113074 330 mm 457 mm Clear Pack of 100
14 options
Product Code. 12300549
Brand: Bryson Packaging™ BAJ360130P
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Transparent and excellent for shipping, storage and as sample bags.
  • Available in a wide range of sizes/capacities
  • With resealable strips
Specifications
Description Resealable Bag
Color Clear
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Width (Metric) 190 mm
Height (Metric) 190 mm