Learn More
Bryson Packaging™ Polyethylene Bags
Description
- Available in a wide range of sizes/capacities
- With resealable strips
Specifications
Specifications
| Description | Resealable Bag |
| Capacity (Metric) | 30 mL |
| Color | Clear |
| For Use With (Application) | Ideal for storing stationery and sending samples |
| Material | Polyethylene |
| Width (Metric) | 40 mm |
| Height (Metric) | 65 mm |