Learn More
Bryson Packaging™ Polyethylene Bags
Product Code. 10113074
Change view
Click to view available options
Width (Metric):
40 mm
60 mm
75 mm
90 mm
100 mm
115 mm
125 mm
150 mm
190 mm
230 mm
255 mm
330 mm
Height (Metric):
65 mm
75 mm
80 mm
115 mm
140 mm
175 mm
190 mm
230 mm
325 mm
355 mm
457 mm
Color:
Clear
Clear with White Bars
Unit Size:
Pack of 100
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) Color Unit Size
10113074 330 mm 457 mm Clear Pack of 100
11398203 40 mm 65 mm Clear Pack of 100
11318213 60 mm 75 mm Clear Pack of 100
11328213 75 mm 80 mm Clear Pack of 100
11978267 90 mm 115 mm Clear Pack of 100
10673995 100 mm 140 mm Clear Pack of 100
11348213 115 mm 115 mm Clear Pack of 100
12320499 125 mm 175 mm Clear Pack of 100
10031474 125 mm 190 mm Clear Pack of 100
10472624 150 mm 230 mm Clear Pack of 100
12300549 190 mm 190 mm Clear Pack of 100
11356934 230 mm 325 mm Clear Pack of 100
11332284 255 mm 355 mm Clear Pack of 100
11388203 330 mm 457 mm Clear with White Bars Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 10113074 Supplier Bryson Packaging™ Supplier No. BAJ360220X
  • ONLINE EXCLUSIVE PRICE
    2420.25 NOK
    251.80 NOK / Pack of 100
    Save 2168.45 NOK
    89% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Transparent and excellent for shipping, storage and as sample bags.
  • Available in a wide range of sizes/capacities
  • With resealable strips
Spezifikation
Description Resealable Bag
Color Clear
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Height (Metric) 457 mm
Width (Metric) 330 mm