Learn More
SPEX CertiPrep™ HG-400 MiniG™ Tissue Homogenizer and Cell Lyser
Shop All SPEX Certiprep Products
missing translation for 'orderingAttributeHoverText'
Packaging Quantity:
1 Ea.
missing translation for 'unitSize'
Each
Description
- Automated mechanical disruption through bead beating,
- Ideal for high-throughput cell lysis applications including DNA/ RNA extraction, protein extraction and pesticide residue analysis
- Window in lid allows to view grinding process
- Lid with safety interlock switch to stops motor if lid is opened during grinding
- LCD screen displays run times
- Specifically designed for rapid cell disruption, cell lysis and tissue homogenization through bead beating
Specifications
Specifications
| Product Type | Tissue Homogenizer |
| Horsepower | 1/7 hp |
| Description | Designed to disrupt cellular materials by oscillating deep-well titer plates vertically |
| For Use With (Application) | Tissue homogenization, DNA/RNA research and extraction, cell lysis, protein extraction, biofuel research, pesticide residue extraction using QuEChERS or the SweEt method. |
| Model | HG-400 MiniG Tissue Homogenizer |
| No. of Strokes | 500 to 1500 strokes/min. |
| Packaging Quantity | 1 Ea. |
| Sample Type | Animal tissue, plant tissue, cell cultures, fruit, cannabis, seeds, yeast and bacteria. |
| Weight (Metric) | 18 kg |