Learn More
Cole-Parmer Miniature Check Valves, Hose Barb, PVDF and Viton™
Product Code. 30054119 Shop All Cole-Parmer Products
Change view
Click to view available options
Inlet:
1/4 in. hose barb
1/8 in. hose barb
3/16 in. hose barb
3/8 in. hose barb
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Inlet unitSize
30054119 1/8 in. hose barb Each
30053824 3/8 in. hose barb Each
30053901 3/16 in. hose barb Each
30053807 1/4 in. hose barb Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30054119 Supplier Cole-Parmer Supplier No. 5011557
  • ONLINE EXCLUSIVE PRICE
    99.30 NOK
    94.45 NOK / Pack of 1
    Save 4.85 NOK
    4% Off
Estimated Shipment: 04-03-2027
Add to basket
Add to basket
Versatile check valves can be installed in any position
Prevent backflow to protect your equipment. Work with both liquids and gases. Use for pressure or vacuum applications. PVDF and Viton construction provides excellent corrosion and chemical resistance. 0.18 psi breaking pressure
Specifications
Tubing Size 0.125 in.
Flow Capacity 0.11
Inlet 1/8 in. hose barb
Outlet Port Barbed
Material (Seal) Viton