Learn More
Cole-Parmer Miniature Check Valves, Hose Barb, Nylon and Silicone
Product Code. 30054057 Shop All Cole-Parmer Products
Change view
Click to view available options
Inlet:
1/4 in. hose barb
1/8 in. hose barb
3/16 in. hose barb
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Inlet unitSize
30054057 1/4 in. hose barb Each
30053887 3/16 in. hose barb Each
30052932 1/8 in. hose barb Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30054057 Supplier Cole-Parmer Supplier No. 5011519
  • ONLINE EXCLUSIVE PRICE
    70.80 NOK
    68.43 NOK / Pack of 1
    Save 2.37 NOK
    3% Off
Estimated Shipment: 25-02-2027
Add to basket
Add to basket
Versatile check valves can be installed in any position
Prevent backflow to protect your equipment. Work with both liquids and gases. Use for pressure or vacuum applications. 0.18 psi breaking pressure
Specifications
Tubing Size 0.25 in.
Flow Capacity 0.13
Inlet 1/4 in. hose barb
Outlet Port Barbed
Material (Seal) Silicone