Learn More
Ekastu Safety™ Eye Wash Bottle with Funnel
Product Code. 10470903
Change view
Click to view available options
Product Type:
Eyewash Bottle
Eyewash Bottle, Empty
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10470903 Eyewash Bottle Each
10385722 Eyewash Bottle, Empty Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10470903 Supplier Ekastu Safety™ Supplier No. 177959
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
Estimated Shipment: 29-12-2026
Made for immediate rinsing of the eyes after accidental splashing from acids, bases and other hazardous chemicals. EKASTU Safety™ Eye Wash Bottle with Funnel can be used in either upright or horizontal positions.
  • Eye Wash Bottle with funnel
  • DIN EN 15154-4
  • Supplied empty or filled
  • For immediate rinsing of the eyes after corrosion by acids, bases and other harsh chemicals
  • PE bottle with screwed-on eye funnel and tilting valve
  • Can be used in both upright and horizontal positions
  • leak-proof
  • Stores easily in EPS Eye Wash Station (sold separately)
Specifications
Capacity (Metric) 600 mL
For Use With (Application) Saline Solution for Immediate Rinsing of the Eyes after Contact with Acids, Bases
Includes Screwed-on Eye Funnel and Tilting Cap
Product Type Eyewash Bottle