Learn More
Trajan Scientific and Medical™ Mini Union Replacement Ferrules
Delivers an easy, leak-free installation for capillary columns without the use of any tools.
Brand:  Trajan Scientific and Medical™ 072697
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13457349
  • 1125.00 NOK / Pack of 10
Estimated Shipment: 14-06-2024
Description
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
Specifications
Specifications
0.5 mm ID
15% Graphite, 85% Vespel
10/Pk.
Bore size: 0.5 mm
Graphite Vespel Replacement Mini Union Ferrule for 0.32mm ID Column
0.32 mm