Learn More
Axygen™ Mini Tube System
Product Code. 12598007 Shop All Axygen Scientific Products
Change view
Click to view available options
Capacity (Metric):
0.65 mL
1.1 mL
12 Cap Strip
8 Cap Strip
Sterility:
Non-sterile
Sterile
Packaging:
12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs.
120 Strips/Bag; 5 Bags/Cs.
120 Strips/PK., 5 Packs/Cs.
8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs.
80 Strips/Bag; 5 Bags/Cs.
80 Strips/PK., 5 Packs/Cs.
96/Rack, 10 Racks/Pk., 5 Packs/Cs.
960 Tubes/Bag, 5 Bags/Cs.
Unit Size:
Pack of 120
Pack of 400
Pack of 4800
Pack of 600
Pack of 80
Pack of 960
Product Code. Capacity (Metric) Sterility Packaging unitSize
12598007 12 Cap Strip Non-sterile 80 Strips/PK., 5 Packs/Cs. Pack of 400
11366324 8 Cap Strip Non-sterile 120 Strips/PK., 5 Packs/Cs. Pack of 120
12529137 8 Cap Strip Non-sterile 120 Strips/PK., 5 Packs/Cs. Pack of 600
11326324 1.1 mL Sterile 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 120
12716829 1.1 mL Sterile 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 600
11346324 12 Cap Strip Non-sterile 80 Strips/PK., 5 Packs/Cs. Pack of 80
11386314 1.1 mL Non-sterile 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 80
12786819 1.1 mL Non-sterile 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 400
10313030 1.1 mL Sterile 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 400
12549207 1.1 mL Sterile 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 80
12746829 1.1 mL Sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 4800
11336324 1.1 mL Sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 960
12539137 8 Cap Strip Sterile 120 Strips/PK., 5 Packs/Cs. Pack of 600
11376324 8 Cap Strip Sterile 120 Strips/PK., 5 Packs/Cs. Pack of 120
13117664 0.65 mL Non-sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 4800
11356314 0.65 mL Non-sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 960
12508017 12 Cap Strip Sterile 80 Strips/PK., 5 Packs/Cs. Pack of 400
11356324 12 Cap Strip Sterile 80 Strips/PK., 5 Packs/Cs. Pack of 80
12776819 0.65 mL Sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 4800
11366314 0.65 mL Sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 960
12588007 1.1 mL Non-sterile 80 Strips/Bag; 5 Bags/Cs. Pack of 400
11376314 1.1 mL Non-sterile 80 Strips/Bag; 5 Bags/Cs. Pack of 80
13121794 1.1 mL Non-sterile 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 600
11316324 1.1 mL Non-sterile 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 120
12736829 1.1 mL Non-sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 4800
12559207 1.1 mL Non-sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 960
12519137 1.1 mL Non-sterile 120 Strips/Bag; 5 Bags/Cs. Pack of 600
11306324 1.1 mL Non-sterile 120 Strips/Bag; 5 Bags/Cs. Pack of 120
12531288 0.65 mL Non-sterile 960 Tubes/Bag, 5 Bags/Cs. Pack of 4800
11346314 0.65 mL Non-sterile 960 Tubes/Bag, 5 Bags/Cs. Pack of 960
11316214 1.1 mL Non-sterile 960 Tubes/Bag, 5 Bags/Cs. Pack of 960
12541288 1.1 mL Non-sterile 960 Tubes/Bag, 5 Bags/Cs. Pack of 4800
32 options
Product Code. 12598007
Brand: Axygen™ MTS12CPC
Only null left
Add to basket
Add to basket
Polypropylene tube system for compound library storage
Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.
  • Rack features a one-way lid design and alpha-numeric grid
  • Molded-in condensation rings on the lid minimize cross contamination
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 12 Cap Strip
Color Clear
Material Polyethylene
Format Strip
For Use With (Application) Ensures positive sample identification
No. per Pack 80/Pk.
Packaging 80 Strips/PK., 5 Packs/Cs.
Sterility Non-sterile
Bottom Shape Strip