Learn More
Axygen™ Mini Tube System
Product Code. 12539137 Shop All Axygen Scientific Products
Click to view available options
Capacity (Metric):
0.65 mL
1.1 mL
12 Cap Strip
8 Cap Strip
Sterility:
Non-sterile
Sterile
Packaging:
12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs.
120 Strips/Bag; 5 Bags/Cs.
120 Strips/PK., 5 Packs/Cs.
8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs.
80 Strips/Bag; 5 Bags/Cs.
80 Strips/PK., 5 Packs/Cs.
96/Rack, 10 Racks/Pk., 5 Packs/Cs.
960 Tubes/Bag, 5 Bags/Cs.
Unit Size:
Pack of 120
Pack of 400
Pack of 4800
Pack of 600
Pack of 80
Pack of 960
Product Code. 12539137
Brand: Axygen™ MTS8CPCS
Only null left
Add to basket
Add to basket
Polypropylene tube system for compound library storage
Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.
  • Rack features a one-way lid design and alpha-numeric grid
  • Molded-in condensation rings on the lid minimize cross contamination
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 8 Cap Strip
Color Clear
Material Polyethylene
Format Strip
For Use With (Application) Ensures positive sample identification
No. per Pack 120/Pk.
Packaging 120 Strips/PK., 5 Packs/Cs.
Sterility Sterile
Bottom Shape Strip