Learn More
Axygen™ Mini Tube System
Product Code. 12508017 Shop All Axygen Scientific Products
Change view
Click to view available options
Capacity (Metric):
0.65 mL
1.1 mL
12 Cap Strip
8 Cap Strip
Sterility:
Non-sterile
Sterile
Packaging:
12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs.
120 Strips/Bag; 5 Bags/Cs.
120 Strips/PK., 5 Packs/Cs.
8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs.
80 Strips/Bag; 5 Bags/Cs.
80 Strips/PK., 5 Packs/Cs.
96/Rack, 10 Racks/Pk., 5 Packs/Cs.
960 Tubes/Bag, 5 Bags/Cs.
Unit Size:
Pack of 120
Pack of 400
Pack of 4800
Pack of 600
Pack of 80
Pack of 960
Product Code. Capacity (Metric) Sterility Packaging unitSize
12508017 12 Cap Strip Sterile 80 Strips/PK., 5 Packs/Cs. Pack of 400
11366324 8 Cap Strip Non-sterile 120 Strips/PK., 5 Packs/Cs. Pack of 120
12529137 8 Cap Strip Non-sterile 120 Strips/PK., 5 Packs/Cs. Pack of 600
11326324 1.1 mL Sterile 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 120
12716829 1.1 mL Sterile 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 600
12598007 12 Cap Strip Non-sterile 80 Strips/PK., 5 Packs/Cs. Pack of 400
11346324 12 Cap Strip Non-sterile 80 Strips/PK., 5 Packs/Cs. Pack of 80
11386314 1.1 mL Non-sterile 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 80
12786819 1.1 mL Non-sterile 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 400
10313030 1.1 mL Sterile 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 400
12549207 1.1 mL Sterile 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 80
12746829 1.1 mL Sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 4800
11336324 1.1 mL Sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 960
12539137 8 Cap Strip Sterile 120 Strips/PK., 5 Packs/Cs. Pack of 600
11376324 8 Cap Strip Sterile 120 Strips/PK., 5 Packs/Cs. Pack of 120
13117664 0.65 mL Non-sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 4800
11356314 0.65 mL Non-sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 960
11356324 12 Cap Strip Sterile 80 Strips/PK., 5 Packs/Cs. Pack of 80
12776819 0.65 mL Sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 4800
11366314 0.65 mL Sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 960
12588007 1.1 mL Non-sterile 80 Strips/Bag; 5 Bags/Cs. Pack of 400
11376314 1.1 mL Non-sterile 80 Strips/Bag; 5 Bags/Cs. Pack of 80
13121794 1.1 mL Non-sterile 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 600
11316324 1.1 mL Non-sterile 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 120
12736829 1.1 mL Non-sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 4800
12559207 1.1 mL Non-sterile 96/Rack, 10 Racks/Pk., 5 Packs/Cs. Pack of 960
12519137 1.1 mL Non-sterile 120 Strips/Bag; 5 Bags/Cs. Pack of 600
11306324 1.1 mL Non-sterile 120 Strips/Bag; 5 Bags/Cs. Pack of 120
12531288 0.65 mL Non-sterile 960 Tubes/Bag, 5 Bags/Cs. Pack of 4800
11346314 0.65 mL Non-sterile 960 Tubes/Bag, 5 Bags/Cs. Pack of 960
11316214 1.1 mL Non-sterile 960 Tubes/Bag, 5 Bags/Cs. Pack of 960
12541288 1.1 mL Non-sterile 960 Tubes/Bag, 5 Bags/Cs. Pack of 4800
32 options
Product Code. 12508017
Brand: Axygen™ MTS12CPCS
Only null left
Add to basket
Add to basket
Polypropylene tube system for compound library storage
Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.
  • Rack features a one-way lid design and alpha-numeric grid
  • Molded-in condensation rings on the lid minimize cross contamination
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 12 Cap Strip
Color Clear
Material Polyethylene
Format Strip
For Use With (Application) Ensures positive sample identification
No. per Pack 80/Pk.
Packaging 80 Strips/PK., 5 Packs/Cs.
Sterility Sterile
Bottom Shape Strip