Learn More
- Home
- Products
- Tubes
- Microtubes
- Axygen™ Mini Tube System
Axygen™ Mini Tube System
Product Code. 11376314 Shop All Axygen Scientific Products| Product Code. | Capacity (Metric) | Sterility | Packaging | unitSize |
|---|---|---|---|---|
| 11376314 | 1.1 mL | Non-sterile | 80 Strips/Bag; 5 Bags/Cs. | Pack of 80 |
| 11366324 | 8 Cap Strip | Non-sterile | 120 Strips/PK., 5 Packs/Cs. | Pack of 120 |
| 12529137 | 8 Cap Strip | Non-sterile | 120 Strips/PK., 5 Packs/Cs. | Pack of 600 |
| 11326324 | 1.1 mL | Sterile | 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 120 |
| 12716829 | 1.1 mL | Sterile | 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 600 |
| 12598007 | 12 Cap Strip | Non-sterile | 80 Strips/PK., 5 Packs/Cs. | Pack of 400 |
| 11346324 | 12 Cap Strip | Non-sterile | 80 Strips/PK., 5 Packs/Cs. | Pack of 80 |
| 11386314 | 1.1 mL | Non-sterile | 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 80 |
| 12786819 | 1.1 mL | Non-sterile | 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 400 |
| 10313030 | 1.1 mL | Sterile | 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 400 |
| 12549207 | 1.1 mL | Sterile | 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 80 |
| 12746829 | 1.1 mL | Sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 4800 |
| 11336324 | 1.1 mL | Sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 960 |
| 12539137 | 8 Cap Strip | Sterile | 120 Strips/PK., 5 Packs/Cs. | Pack of 600 |
| 11376324 | 8 Cap Strip | Sterile | 120 Strips/PK., 5 Packs/Cs. | Pack of 120 |
| 13117664 | 0.65 mL | Non-sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 4800 |
| 11356314 | 0.65 mL | Non-sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 960 |
| 12508017 | 12 Cap Strip | Sterile | 80 Strips/PK., 5 Packs/Cs. | Pack of 400 |
| 11356324 | 12 Cap Strip | Sterile | 80 Strips/PK., 5 Packs/Cs. | Pack of 80 |
| 12776819 | 0.65 mL | Sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 4800 |
| 11366314 | 0.65 mL | Sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 960 |
| 12588007 | 1.1 mL | Non-sterile | 80 Strips/Bag; 5 Bags/Cs. | Pack of 400 |
| 13121794 | 1.1 mL | Non-sterile | 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 600 |
| 11316324 | 1.1 mL | Non-sterile | 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 120 |
| 12736829 | 1.1 mL | Non-sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 4800 |
| 12559207 | 1.1 mL | Non-sterile | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Pack of 960 |
| 12519137 | 1.1 mL | Non-sterile | 120 Strips/Bag; 5 Bags/Cs. | Pack of 600 |
| 11306324 | 1.1 mL | Non-sterile | 120 Strips/Bag; 5 Bags/Cs. | Pack of 120 |
| 12531288 | 0.65 mL | Non-sterile | 960 Tubes/Bag, 5 Bags/Cs. | Pack of 4800 |
| 11346314 | 0.65 mL | Non-sterile | 960 Tubes/Bag, 5 Bags/Cs. | Pack of 960 |
| 11316214 | 1.1 mL | Non-sterile | 960 Tubes/Bag, 5 Bags/Cs. | Pack of 960 |
| 12541288 | 1.1 mL | Non-sterile | 960 Tubes/Bag, 5 Bags/Cs. | Pack of 4800 |
Product Code. 11376314
Brand: Axygen™ MTS1112C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.
- Rack features a one-way lid design and alpha-numeric grid
- Molded-in condensation rings on the lid minimize cross contamination
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 1.1 mL |
| Color | Clear |
| Material | Polypropylene |
| Format | Bag |
| For Use With (Application) | Ensures positive sample identification |
| Includes | Alpha numeric grid, one way lid |
| No. per Pack | 80/Pk. |
| Packaging | 80 Strips/Bag; 5 Bags/Cs. |
| Sterility | Non-sterile |
| Graduated | Yes |
| Bottom Shape | Round |
| Product Type | Microtube |
| Show More Show Less |