Learn More
Brady™ Mini Lockout Starter Kit
Product Code. 11726795 Shop All Brady Products
Change view
Click to view available options
Color:
Black/Blue/Orange/Red
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11726795 Black/Blue/Orange/Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11726795 Supplier Brady™ Supplier No. 805856
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
In Stock
The Mini Lockout Starter Kit is an economical kit which will allow you to lockout a variety of equipment.
  • An economical kit which will allow you to lockout a variety of equipment.
Specifications
Product Type Mini Lockout Starter Kit
Color Black/Blue/Orange/Red
For Use With (Application) Lockout Tagout
Quantity 1 Ea.