Learn More
Invitrogen™ Mini Gel Tank Base
Product Code. 15344574
Change view
Click to view available options
Quantity:
1 unit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15344574 1 unit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15344574 Supplier Invitrogen™ Supplier No. A25950
The Mini Gel Tank Base is a replacement part for the Mini Gel Tank. It holds the Gel Runner Tank.
About the Mini Gel Tank
The Mini Gel Tank is compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.
For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Mini Gels
Gel Compatibility Novex™ Mini Gels
Gel Size Mini
Type Mini Gel Tank Base
Quantity 1 unit
Shipping Condition Room Temperature