Learn More
Thermo Scientific™ Mini Capillary Unions
Product Code. 10311155 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
0.22 to 0.32 mm ID Capillary GC Column
0.53 mm ID Capillary GC Column
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10311155 0.22 to 0.32 mm ID Capillary GC Column Each
10289074 0.53 mm ID Capillary GC Column Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10311155 Supplier Thermo Scientific™ Supplier No. 290GU498
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Kit contains one union and five ferrules
Includes:
Union, 1; Ferrules, 5
TRUSTED_SUSTAINABILITY
Specifications
Type Capillary Union
Description For 0.32 mm I.D. Capillary
For Use With (Equipment) 0.22 to 0.32 mm ID Capillary GC Column