missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil, Light Weight, MP Biomedicals

Product Code. 11486581 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
Unit Size:
25mL
500mL
Product Code. Quantity unitSize
11486581 25 mL 25mL
11496581 500 mL 500mL
2 options
This item is not returnable. View return policy

Product Code. 11486581

Brand: MP Biomedicals 0219483625

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number MFCD00131611
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Density 0.81 to 0.894g/mL
Quantity 25 mL
Chemical Name or Material Mineral Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.