missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MINA Rabbit anti-Human, Mouse, Rat, Clone: 4E6G6, Novus Biologicals™
Rabbit Monoclonal Antibody
1760.00 NOK - 4590.00 NOK
Specifications
| Antigen | MINA |
|---|---|
| Clone | 4E6G6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18324536
|
Novus Biologicals
NBP3-15931-20UL |
20 μg |
1760.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18324753
|
Novus Biologicals
NBP3-15931-100UL |
100 μg |
4590.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MINA Monoclonal antibody specifically detects MINA in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MINA | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 84864 | |
| IgG | |
| Affinity purified |
| 4E6G6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp762O1912, FLJ14393, MDIGDKFZp762O1912, MINA53mdig, mineral dust induced gene protein, Mineral dust-induced gene protein, MYC induced nuclear antigen, MYC-induced nuclear antigen, myc-induced nuclear antigen, 53 kDa, NO52, Nucleolar protein 52 | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human MINA (Q8IUF8). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title