Learn More
Cole-Parmer™ Miltex™ Iris Tissue Forceps
Product Code. 11760309 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric):
10.16 cm
Unit Size:
Each
Product Code. Length (Metric) unitSize
11760309 10.16 cm Each
1 options
Product Code. 11760309
Brand: Cole-Parmer™ 1092302
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Feature the highest quality stainless steel, crafted to the most exacting standards
  • Precision cutting and holding surfaces
  • Ratcheting lock mechanisms
  • Non-glare finish
Specifications
Tip Style Straight
Length (English) 4 in.
Material Stainless Steel
Length (Metric) 10.16 cm
Product Type Precision-grade Iris Tissue Forceps
Type Iris Tissue Forceps