Learn More
Milnacipran ((1S-cis) hydrochloride), MedChemExpress
Product Code. 16484267
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16484267 50 mg 50mg
16478378 10 mg 10mg
16450798 5 mg 5mg
3 options
Product Code. 16484267
Brand: Medchem Express HYB0168B
MedChemExpress Milnacipran (1S-cis) hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI), used in the clinical treatment of fibromyalgia.
Milnacipran (1S-cis) hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI), used in the clinical treatment of fibromyalgia.
Specifications
Chemical Name or Material Milnacipran ((1S-cis) hydrochloride)
Physical Form Solid
CAS 175131-60-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H23ClN2O
For Use With (Application) Neuroscience-Neuromodulation
Synonym Levomilnacipran hydrochloride F-2695 hydrochloride
Solubility Information H2O : ≥ 50 mg/mL (176.80 mM)
SMILES O=C([C@]1(C2=CC=CC=C2)[C@H](CN)C1)N(CC)CC.[H]Cl
Molecular Weight (g/mol) 282.81
Formula Weight 282.81
Percent Purity 99.94%
Grade Research