missing translation for 'onlineSavingsMsg'
Learn More

Milnacipran ((1S-cis) hydrochloride), MedChemExpress

Product Code. 16478378
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16478378

Brand: Medchem Express HYB0168B

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Milnacipran (1S-cis) hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI), used in the clinical treatment of fibromyalgia.

Milnacipran (1S-cis) hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI), used in the clinical treatment of fibromyalgia.

Specifications

Chemical Name or Material Milnacipran ((1S-cis) hydrochloride)
Physical Form Solid
CAS 175131-60-9
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H23ClN2O
For Use With (Application) Neuroscience-Neuromodulation
Synonym Levomilnacipran hydrochloride F-2695 hydrochloride
Solubility Information H2O : ≥ 50 mg/mL (176.80 mM)
SMILES O=C([C@]1(C2=CC=CC=C2)[C@H](CN)C1)N(CC)CC.[H]Cl
Molecular Weight (g/mol) 282.81
Formula Weight 282.81
Percent Purity 99.94%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.