Learn More
IKA™ Mills MultiDrive control MT Package, EU-Plug
Product Code. 17373610 Shop All IKA Products
Change view
Click to view available options
Capacity (Metric):
150 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17373610 150 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17373610 Supplier IKA™ Supplier No. 0010007083
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
In Stock
The Mills MultiDrive control MT Package can perform a wide range of crushing tasks involving coarse and fine crushing without cross-contamination, thanks to the variety of vessels available.
  • Crushing without cross-contamination
    • This is possible with the MultiDrive control package including autoclavable disposable tubes MT 150 and the required TC 1 protective housing for dry samples.
  • High performance
    • Excellent crushing performance is guaranteed by a combination of variable rotational speeds, ranging from 3000 RPM to 20,000 RPM, and a 1000 Watt output.
  • Interval operation
    • The option of interval programming is simple to activate at the press of a button. Interval operation is an asset during the coarse crushing of hard samples or for extra thorough blending.
  • Integrated cooling
    • A cooling system is integrated in the milling cup, which allows indirect heat dissipation. Thus, coolant and sample remain separate.
  • TFT-display
    • A clear TFT-display guarantees user-friendly operation.
  • Temperature measurement
    • MultiDrive control offers temperature measurement and vessel recognition using RFID. A temperature limit can be set for temperature-sensitive materials, or for specific planned reactions to protect against overheating.
  • Weighing function
    • The weighing function is also integrated into the user-friendly workflow of the IKA MultiDrive control. Weighing is performed before crushing using the same mixing cup, so that no transfer of contents is required.
  • Interface
    • There is a USB interface available for easy actuation and documentation respectively.
  • Scope of delivery
    • IKA MultiDrive control
    • MultiDrive TC 1IKA
    • MultiDrive MT 150.5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Universal Blender/Mill
Autoclavable Autoclavable
Capacity (Metric) 150 mL
Display Type TFT-display
Disposable Single-use
For Use With (Application) Milling of dry samples
Hertz 50/60 Hz
Includes IKA MultiDrive control, MultiDrive TC 1 protective housing, 5 IKA MultiDrive MT 150.5 milling tubes
Interface USB, Bluetooth
IP Rating IP31
Plug Type EU plug
Speed 3000 to 20,000 rpm
Voltage 220/240 V
Watts 1000 W
Weight (Metric) 8.5 kg