Learn More
IKA™ Mills MultiDrive control BT Package, EU-Plug
Product Code. 17303620 Shop All IKA Products
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17303620 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17303620 Supplier IKA™ Supplier No. 0010007086
  • ONLINE EXCLUSIVE PRICE
    52840.00 NOK
    48983.37 NOK / Pack of 1
    Save 3856.63 NOK
    7% Off
View all promotions
In Stock
The Mills MultiDrive control BT Package crushes without cross-contamination using autoclavable disposable tubes BT 250.
BT 250 is a disposable tube with a cross-shaped stainless steel beater and flow breakers. The plastic blending tube can be autoclaved. This effectively prevents cross-contamination even if it is used multiple times. An integrated RFID chip simultaneously stores and limits the lifespan of the blending tube.
  • Crushing without cross-contamination
    • This is possible with the MultiDrive control package including autoclavable disposable tubes BT 250 and the required TC 1 protective housing for dry samples.
  • High performance
    • Excellent crushing performance is guaranteed by a combination of variable rotational speeds, ranging from 3000 RPM to 20,000 RPM, and a 1000 Watt output.
  • Interval operation
    • The option of interval programming is simple to activate at the press of a button. Interval operation is an asset during the coarse crushing of hard samples or for extra thorough blending.
  • Integrated cooling
    • A cooling system is integrated in the milling cup, which allows indirect heat dissipation. Thus, coolant and sample remain separate.
  • TFT-display
    • A clear TFT-display guarantees user-friendly operation.
  • Temperature measurement
    • MultiDrive control offers temperature measurement and vessel recognition using RFID. A temperature limit can be set for temperature-sensitive materials, or for specific planned reactions to protect against overheating.
  • Weighing function
    • The weighing function is also integrated into the user-friendly workflow of the IKA MultiDrive control. Weighing is performed before crushing using the same mixing cup, so that no transfer of contents is required.
  • Scope of delivery
    • IKA MultiDrive control
    • MultiDrive TC 1 IKA
    • MultiDrive BT 250.5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Universal Blender/Mill
Autoclavable Autoclavable
Capacity (Metric) 250 mL
Display Type TFT-display
Disposable Single-use
For Use With (Application) Milling of samples together with liquid
Hertz 50/60 Hz
Includes IKA MultiDrive control, MultiDrive TC 1 protective housing, 5 IKA MultiDrive BT 250.5 blending tubes
Interface USB, Bluetooth
IP Rating IP31
Plug Type EU plug
Speed 3000 to 20,000 rpm
Voltage 230 V
Watts 1000 W
Weight (Metric) 8.5 kg