Learn More
Merck Millicell Hanging Cell Culture Insert
Shop All Merck Products
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
5 μm
8 μm
No. of Wells:
6
12
24
Unit Size:
Pack of 48
Description
- Cell growth, structure, and function more closely mimic what occurs in vivo
- In addition, Millicell inserts make it possible to study both sides of the cell monolayer
- The inserts are easily prepared for SEM and TEM visualizing techniques, and they are compatible with cellular and/or fluorescent stains
- For co-culturing and permeability assays
- Unique design allows easier basolateral access than other hanging inserts with less risk of contamination
- Available in 5 pore sizes and 3 diameters, including a 1 μm pore size that is optically transparent for better visualization by microscopy
- PET Membrane (polyethylene terephthalate) for growth of attachment-dependent cells without matrix
- Individually blister packaged
Specifications
Specifications
| Description | PET (Polyethylene Terephthalate) Filter Code |
| Material | Polystyrene |
| Pore Size | 5 μm |
| Type | Cell Culture Insert |
| Item Description | 50°C Max. Operating Temperature |
| For Use With (Application) | Cell Attachment, Cell Culture, Cell Differentiation, Immunocytochemistry |
| Diameter (Metric) Filter | 24 mm |
| Quantity | 48/Pk |
| For Use With | 6-well Plates |
| Color | Clear |
| Show More |