Learn More
Honeywell Safety Products™ Millenia Safety Glasses
Click to view available options
Frame Color:
Black
Blue
Silver
Lens Tint:
Clear
Silver Mirror
Yellow
Includes:
Adjustable, breakaway Flexicord retainer
Flexicord retainer
Lens Coating:
Impact-resistant, Anti-fog, UV-treated
Impact-resistant, Scratch-resistant, UV-treated
Unit Size:
Each
Description
Plano lens fashion safety spectacle, wrap around design with abrasion resistant lens and integral side shields. Available in a range of frame and lens colors.
Specifications
Specifications
| Certifications/Compliance | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1131, EC Attestation Number EC1131 |
| Lens Coating | Impact-resistant, Anti-fog, UV-treated |
| Lens Tint | Clear |
| Product Type | Safety Glasses |
| For Use With (Application) | Administration, Automotive, Energy or Electricity, Laboratory, Medical and Pharmaceutical and Wood industries |
| Frame Color | Blue |
| Includes | Flexicord retainer |
| No. per Pack | 1 Ea. |
| Weight (Metric) | 32 g |
| Lens Material | Polycarbonate |
| Show More |